siRNA Details
| Gene Symbol: Zdbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267872.1 | NP_001254801.1 | 6 | 849 | ||
| NM_001285936.1 | NP_001272865.1 | 7 | 983 | |||
| NM_001285937.1 | NP_001272866.1 | 6 | 926 | |||
| NM_028673.1 | NP_082949.1 | 7 | 899 | |||
| GenBank mRNA | AB777269.1 | - | 6 | 926 | ||
| AB777270.1 | - | 7 | 983 | |||
| AK015271.1 | - | 7 | 900 | |||
| AK132808.1 | - | 6 | 850 | |||
| BC050985.1 | - | 460 | ||||

Zdbf2
Genomic Map