siRNA Details
| Gene Symbol: Arhgap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176837.2 | NP_789807.1 | 2 | 255 | ||
| GenBank mRNA | AK014728.1 | - | 2 | 208 | ||
| AK087800.1 | - | 2 | 231 | |||
| BC030858.1 | - | 2 | 255 | 4194996 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap18| Gene Symbol: Arhgap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176837.2 | NP_789807.1 | 2 | 255 | ||
| GenBank mRNA | AK014728.1 | - | 2 | 208 | ||
| AK087800.1 | - | 2 | 231 | |||
| BC030858.1 | - | 2 | 255 | 4194996 | ||