siRNA Details
| Gene Symbol: Lyrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285959.1 | NP_001272888.1 | 2 | 136 | ||
| NM_001285960.1 | NP_001272889.1 | 2 | 269 | |||
| NM_029610.3 | NP_083886.1 | 2 | 520 | |||
| GenBank mRNA | AK009135.1 | - | 2 | 523 | ||
| AK015083.1 | - | 3 | 328 | |||
| AK157931.1 | - | 138 | ||||
| BC011339.1 | - | 2 | 310 | 3978191 | ||
| BY752674.1 | - | 2 | 138 | |||
| DV043140.1 | - | 1 | 294 | |||

Lyrm1
Genomic Map