siRNA Details
| Gene Symbol: Ccdc175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028687.1 | NP_082963.1 | 4 | 464 | ||
| GenBank mRNA | AK015067.1 | - | 4 | 453 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc175| Gene Symbol: Ccdc175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028687.1 | NP_082963.1 | 4 | 464 | ||
| GenBank mRNA | AK015067.1 | - | 4 | 453 | ||