siRNA Details
| Gene Symbol: Rftn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028713.1 | NP_082989.1 | 5 | 1379 | ||
| GenBank mRNA | AK012229.2 | - | 3 | 1074 | ||
| AK014318.1 | - | 5 | 1131 | |||
| BC026871.1 | - | 5 | 943 | 3966594 | ||
| BC038341.1 | - | 5 | 1379 | 4218473 | ||

Rftn2
Genomic Map