siRNA Details
| Gene Symbol: Rftn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028713.1 | NP_082989.1 | 3, 4 | 960 | ||
| GenBank mRNA | AK014318.1 | - | 3, 4 | 712 | ||
| BC026871.1 | - | 3, 4 | 524 | 3966594 | ||
| BC038341.1 | - | 3, 4 | 960 | 4218473 | ||

Rftn2
Genomic Map