siRNA Details
| Gene Symbol: Rd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB030195.1 | - | 3 | 1184 | ||
| AK014383.2 | - | 3 | 1060 | |||
| AK045045.1 | - | 3 | 1194 | |||
| BC017602.1 | - | 3 | 1136 | 4503004 | ||
| BC089516.1 | - | 3 | 1346 | |||
| BC103778.1 | - | 3 | 1128 | 6493935 | ||

Rd3
Genomic Map