siRNA Details
| Gene Symbol: Rin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028724.4 | NP_083000.5 | 13 | 2704 | ||
| GenBank mRNA | AK014548.1 | - | 5 | 1176 | ||
| AK040763.1 | - | 5 | 1176 | |||
| AK150680.1 | - | 11 | 2512 | |||
| AK151332.1 | - | 11 | 2468 | |||
| AK152025.1 | - | 6 | 1149 | |||
| AK152893.1 | - | 11 | 2512 | |||
| AK154421.1 | - | 5 | 1120 | |||
| AK163047.1 | - | 5 | 1691 | |||
| AK171064.1 | - | 5 | 1091 | |||
| BC005529.1 | - | 5 | 1137 | 3495681 | ||
| BC040390.1 | - | 13 | 2698 | 3156682 | ||

Rin2
Genomic Map