siRNA Details
| Gene Symbol: Ddias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080995.1 | NP_001074464.1 | 7 | 2461 | ||
| GenBank mRNA | AK014608.2 | - | 6 | 2390 | ||
| AK154915.1 | - | 6 | 2366 | |||
| BC066148.1 | - | 6 | 2352 | 30086532 | ||
| DQ400346.1 | - | 6 | 2392 | |||

Ddias
Genomic Map