siRNA Details
| Gene Symbol: Ddias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080995.1 | NP_001074464.1 | 7 | 1769 | ||
| GenBank mRNA | AK154915.1 | - | 6 | 1674 | ||
| BC066148.1 | - | 6 | 1660 | 30086532 | ||
| DQ400346.1 | - | 6 | 1700 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddias| Gene Symbol: Ddias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080995.1 | NP_001074464.1 | 7 | 1769 | ||
| GenBank mRNA | AK154915.1 | - | 6 | 1674 | ||
| BC066148.1 | - | 6 | 1660 | 30086532 | ||
| DQ400346.1 | - | 6 | 1700 | |||