siRNA Details
| Gene Symbol: 4921501E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009544.2 | NP_001009544.1 | 1 | 2050 | ||
| GenBank mRNA | AK014800.1 | - | 1 | 1998 | ||
| AK077044.1 | - | 1 | 2018 | |||
| BC150955.1 | - | 1 | 1897 | 9087866 | ||

4921501E09Rik
Genomic Map