siRNA Details
| Gene Symbol: Steap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103156.2 | NP_001096626.1 | 4 | 1028 | ||
| NM_001103157.2 | NP_001096627.1 | 4 | 1383 | |||
| NM_001285469.1 | NP_001272398.1 | 4 | 937 | |||
| NM_001285470.1 | NP_001272399.1 | 3 | 902 | |||
| NM_001285471.1 | NP_001272400.1 | 3 | 902 | |||
| NM_028734.5 | NP_083010.2 | 4 | 1085 | |||
| GenBank mRNA | AK052981.1 | - | 4 | 1087 | ||
| AK086787.1 | - | 4 | 1056 | |||
| AK089927.1 | - | 3 | 876 | |||
| AK145468.1 | - | 4 | 974 | |||
| AK162343.1 | - | 3 | 864 | |||
| AK165644.1 | - | 4 | 1348 | |||
| AK166162.1 | - | 4 | 1052 | |||
| AK166607.1 | - | 4 | 974 | |||
| BC043459.1 | - | 4 | 938 | |||
| BC150881.1 | - | 4 | 944 | 9087792 | ||
| BC150889.1 | - | 4 | 944 | 9087800 | ||
| BC150997.1 | - | 995 | 9087910 | |||
| BC151008.1 | - | 995 | 9087921 | |||

Steap2
Genomic Map