siRNA Details
| Gene Symbol: Ttc21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028735.3 | NP_083011.2 | 3, 4 | 389 | ||
| GenBank mRNA | AK015017.1 | - | 3, 4 | 349 | ||
| AK029952.1 | - | 3, 4 | 393 | |||
| AK133304.1 | - | 3, 4 | 361 | |||
| AK163163.1 | - | 3, 4 | 360 | |||
| BC133699.1 | - | 3, 4 | 347 | 40098338 | ||
| DQ011665.1 | - | 3, 4 | 393 | |||

Ttc21a
Genomic Map