siRNA Details
| Gene Symbol: Ttc21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028735.3 | NP_083011.2 | 28 | 3936 | ||
| GenBank mRNA | AK015017.1 | - | 28 | 3896 | ||
| AK029952.1 | - | 28 | 3940 | |||
| BC133699.1 | - | 28 | 3894 | 40098338 | ||
| DQ011665.1 | - | 28 | 3940 | |||

Ttc21a
Genomic Map