siRNA Details
| Gene Symbol: Asz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023729.3 | NP_076218.3 | 6 | 683 | ||
| GenBank mRNA | AF459789.1 | - | 6 | 677 | ||
| AK016595.1 | - | 6 | 678 | |||
| BC058859.1 | - | 6 | 692 | 30011651 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asz1| Gene Symbol: Asz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023729.3 | NP_076218.3 | 6 | 683 | ||
| GenBank mRNA | AF459789.1 | - | 6 | 677 | ||
| AK016595.1 | - | 6 | 678 | |||
| BC058859.1 | - | 6 | 692 | 30011651 | ||