siRNA Details
| Gene Symbol: Npl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028749.1 | NP_083025.1 | 8 | 655 | ||
| GenBank mRNA | AK002734.1 | - | 8 | 655 | ||
| AK088859.1 | - | 8 | 657 | |||
| BC022734.1 | - | 8 | 651 | 4506776 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npl| Gene Symbol: Npl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028749.1 | NP_083025.1 | 8 | 655 | ||
| GenBank mRNA | AK002734.1 | - | 8 | 655 | ||
| AK088859.1 | - | 8 | 657 | |||
| BC022734.1 | - | 8 | 651 | 4506776 | ||