siRNA Details
| Gene Symbol: MYO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012334.2 | NP_036466.2 | 36 | 5480 | ||
| XM_005248306.4 | XP_005248363.1 | 18 | 4374 | |||
| XM_005248307.2 | XP_005248364.1 | 18 | 3850 | |||
| XM_006714475.2 | XP_006714538.1 | 35 | 5187 | |||
| XM_011514046.2 | XP_011512348.1 | 18 | 3553 | |||
| GenBank mRNA | AB018342.2 | - | 36 | 5467 | ||
| AF132022.1 | - | 3 | 421 | |||
| AF234532.1 | - | 36 | 5234 | |||
| AF247457.2 | - | 36 | 5165 | |||
| AK001501.1 | - | 2 | 128 | |||
| AK074717.1 | - | 12 | 1826 | |||
| AK295630.1 | - | 11 | 1728 | |||
| AK295736.1 | - | 18 | 3335 | |||
| AK303459.1 | - | 6 | 1074 | |||
| AK316138.1 | - | 18 | 3508 | |||
| AL832428.1 | - | 26 | 3934 | |||
| BC137168.1 | - | 36 | 5189 | 9021165 | ||
| BC150285.1 | - | 36 | 5467 | 8860110 | ||
| CA488271.1 | - | 2 | 323 | |||

MYO10
Genomic Map