siRNA Details
| Gene Symbol: Gga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028758.2 | NP_083034.1 | 7 | 753 | ||
| GenBank mRNA | AK004632.1 | - | 7 | 725 | ||
| AK034612.1 | - | 4 | 396 | |||
| AK129280.1 | - | 7 | 712 | |||
| AK143691.1 | - | 7 | 741 | |||
| AK149798.1 | - | 7 | 743 | |||
| AK154904.1 | - | 7 | 728 | |||
| AK159953.1 | - | 7 | 743 | |||
| BC062933.1 | - | 7 | 722 | 30356060 | ||

Gga2
Genomic Map