siRNA Details
| Gene Symbol: Gga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028758.2 | NP_083034.1 | 17 | 1867 | ||
| GenBank mRNA | AK004632.1 | - | 17 | 1839 | ||
| AK034612.1 | - | 14 | 1510 | |||
| AK129280.1 | - | 17 | 1826 | |||
| AK143691.1 | - | 17 | 1855 | |||
| AK149798.1 | - | 17 | 1857 | |||
| AK154904.1 | - | 17 | 1842 | |||
| AK159953.1 | - | 17 | 1857 | |||
| AK180517.1 | - | 2 | 145 | |||
| BC057377.1 | - | 10 | 1370 | 5698559 | ||
| BC062933.1 | - | 17 | 1836 | 30356060 | ||

Gga2
Genomic Map