siRNA Details
| Gene Symbol: Cep55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164362.1 | NP_001157834.1 | 6, 7 | 1119 | ||
| NM_028293.1 | NP_082569.1 | 5, 6 | 743 | |||
| NM_028760.2 | NP_083036.2 | 6, 7 | 1095 | |||
| GenBank mRNA | AK004655.1 | - | 6, 7 | 1121 | ||
| AK028216.1 | - | 5, 6 | 729 | |||
| AK088548.1 | - | 6, 7 | 802 | |||
| BC026966.1 | - | 3, 4 | 451 | |||
| BC031396.1 | - | 6, 7 | 1048 | 4950530 | ||

Cep55
Genomic Map