siRNA Details
| Gene Symbol: Rbm19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028762.1 | NP_083038.1 | 21 | 2531 | ||
| GenBank mRNA | AK004657.1 | - | 12 | 1346 | ||
| AK146493.1 | - | 21 | 2564 | |||
| AK167100.1 | - | 21 | 2528 | |||
| AK216411.1 | - | 2 | 78 | |||
| BC025619.1 | - | 21 | 2531 | 5320947 | ||
| BC034010.1 | - | 21 | 2497 | 4225239 | ||

Rbm19
Genomic Map