siRNA Details
| Gene Symbol: Crot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023733.3 | NP_076222.1 | 3, 4 | 281 | ||
| GenBank mRNA | AK004567.2 | - | 3, 4 | 282 | ||
| AK037973.1 | - | 2, 3 | 733 | |||
| BC006593.1 | - | 3, 4 | 269 | 3156501 | ||
| BC012308.1 | - | 3, 4 | 296 | 3500167 | ||

Crot
Genomic Map