siRNA Details
| Gene Symbol: Pi16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023734.3 | NP_076223.3 | 5 | 1068 | ||
| GenBank mRNA | AI587910.1 | - | 3 | 347 | ||
| AK004672.1 | - | 4 | 1247 | |||
| AK157854.1 | - | 4 | 1559 | |||
| AK158339.1 | - | 4 | 1555 | |||
| BC039124.1 | - | 5 | 665 | |||

Pi16
Genomic Map