siRNA Details
| Gene Symbol: Zfp263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148924.3 | NP_683726.2 | 4 | 1012 | ||
| GenBank mRNA | AF499776.1 | - | 4 | 980 | ||
| AK004765.2 | - | 4 | 1009 | |||
| AK159455.1 | - | 4 | 1000 | |||
| AK182706.1 | - | 4 | 366 | |||
| BC011505.1 | - | 5 | 1538 | |||
| BC056222.1 | - | 4 | 833 | 30009326 | ||
| BC060162.1 | - | 3 | 1945 | |||

Zfp263
Genomic Map