siRNA Details
| Gene Symbol: Zfp263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148924.3 | NP_683726.2 | 6 | 2079 | ||
| GenBank mRNA | AF499776.1 | - | 6 | 2047 | ||
| AK004765.2 | - | 6 | 2076 | |||
| AK159455.1 | - | 6 | 2067 | |||
| BC011505.1 | - | 7 | 2605 | |||
| BC056222.1 | - | 6 | 1900 | 30009326 | ||
| BC060162.1 | - | 5 | 3012 | |||

Zfp263
Genomic Map