siRNA Details
| Gene Symbol: Krt80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028770.2 | NP_083046.2 | 5 | 887 | ||
| GenBank mRNA | AK004811.2 | - | 4 | 676 | ||
| AK171114.1 | - | 776 | ||||
| BC119366.1 | - | 5 | 887 | 8734115 | ||
| BC120601.1 | - | 5 | 887 | 40129524 | ||

Krt80
Genomic Map