siRNA Details
| Gene Symbol: Smg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024262.1 | NP_077224.1 | 1 | 692 | ||
| GenBank mRNA | AK004714.1 | - | 1 | 685 | ||
| BC020005.1 | - | 1 | 692 | 3981459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smg8| Gene Symbol: Smg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024262.1 | NP_077224.1 | 1 | 692 | ||
| GenBank mRNA | AK004714.1 | - | 1 | 685 | ||
| BC020005.1 | - | 1 | 692 | 3981459 | ||