siRNA Details
| Gene Symbol: Smg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024262.1 | NP_077224.1 | 3 | 1961 | ||
| GenBank mRNA | AK004714.1 | - | 3 | 1954 | ||
| AK031106.1 | - | 3 | 1935 | |||
| AK035089.1 | - | 3 | 928 | |||
| BC002247.1 | - | 3 | 566 | |||
| BC020005.1 | - | 3 | 1961 | 3981459 | ||

Smg8
Genomic Map