siRNA Details
| Gene Symbol: Opa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199177.1 | NP_001186106.1 | 22, 23 | 2423 | ||
| NM_133752.3 | NP_598513.1 | 21, 22 | 2369 | |||
| GenBank mRNA | AB044138.1 | - | 21, 22 | 2361 | ||
| AK004715.1 | - | 22, 23 | 2379 | |||
| AK029157.1 | - | 21, 22 | 2327 | |||
| AK030835.1 | - | 22, 23 | 2398 | |||
| AK122311.1 | - | 21, 22 | 2298 | |||
| AK145620.1 | - | 21, 22 | 2321 | |||
| BC025160.1 | - | 3, 4 | 385 | |||
| BC052704.1 | - | 20, 21 | 3161 | |||
| BC138665.1 | - | 21, 22 | 2250 | 8861686 | ||
| BC145959.1 | - | 21, 22 | 2250 | 40131262 | ||

Opa1
Genomic Map