siRNA Details
| Gene Symbol: Robo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF536772.1 | - | 15, 16 | 2509 | ||
| AK004723.2 | - | 15, 16 | 2485 | |||
| AK030973.1 | - | 1276 | ||||
| AK087355.1 | - | 14, 15 | 2659 | |||
| AK172104.1 | - | 14, 15 | 2747 | |||
| BC020129.1 | - | 7, 8 | 1029 | |||
| BC071193.1 | - | 14, 15 | 2199 | 5060473 | ||

Robo4
Genomic Map