siRNA Details
| Gene Symbol: Robo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF536772.1 | - | 9, 10 | 1549 | ||
| AK004723.2 | - | 9, 10 | 1546 | |||
| AK030973.1 | - | 336 | ||||
| AK087355.1 | - | 8, 9 | 1699 | |||
| AK172104.1 | - | 8, 9 | 1787 | |||
| BC020129.1 | - | 1, 2 | 90 | |||
| BC071193.1 | - | 8, 9 | 1239 | 5060473 | ||

Robo4
Genomic Map