siRNA Details
| Gene Symbol: Zfp946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198003.2 | NP_932120.1 | 7 | 1007 | ||
| GenBank mRNA | AK004870.2 | - | 7 | 1025 | ||
| BC025651.1 | - | 7 | 1071 | 5343080 | ||
| BC098460.1 | - | 7 | 1016 | 30061104 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp946| Gene Symbol: Zfp946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198003.2 | NP_932120.1 | 7 | 1007 | ||
| GenBank mRNA | AK004870.2 | - | 7 | 1025 | ||
| BC025651.1 | - | 7 | 1071 | 5343080 | ||
| BC098460.1 | - | 7 | 1016 | 30061104 | ||