siRNA Details
| Gene Symbol: Stra6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004855.1 | - | 3, 4 | 304 | ||
| AK045033.1 | - | 4, 5 | 732 | |||
| AK050152.1 | - | 3, 4 | 315 | |||
| AK050348.1 | - | 3, 4 | 311 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stra6l| Gene Symbol: Stra6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004855.1 | - | 3, 4 | 304 | ||
| AK045033.1 | - | 4, 5 | 732 | |||
| AK050152.1 | - | 3, 4 | 315 | |||
| AK050348.1 | - | 3, 4 | 311 | |||