siRNA Details
| Gene Symbol: Stra6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004855.1 | - | 3 | 228 | ||
| AK050152.1 | - | 3 | 239 | |||
| AK050348.1 | - | 3 | 235 | |||
| BC111889.1 | - | 3 | 165 | 40060643 | ||
| BC112408.1 | - | 3 | 165 | 40060646 | ||
| BY776586.1 | - | 4 | 315 | |||

Stra6l
Genomic Map