siRNA Details
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197024.1 | NP_001183953.1 | 11 | 1316 | ||
| NM_001290736.1 | NP_001277665.1 | 11 | 1327 | |||
| NM_028789.3 | NP_083065.1 | 2 | 183 | |||
| GenBank mRNA | AK004898.1 | - | 2 | 184 | ||
| BC059910.1 | - | 11 | 1327 | |||
| BC089378.1 | - | 2 | 69 | |||
| BC150842.1 | - | 2 | 108 | 9087753 | ||

Unkl
Genomic Map