siRNA Details
| Gene Symbol: Josd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028792.3 | NP_083068.1 | 2 | 1191 | ||
| GenBank mRNA | AK004913.1 | - | 2 | 1140 | ||
| AK154799.1 | - | 2 | 1240 | |||
| AK211745.1 | - | 2 | 317 | |||
| AK214040.1 | - | 1 | 55 | |||
| BC006928.1 | - | 2 | 339 | 2655826 | ||
| BC086769.1 | - | 2 | 311 | 5707990 | ||

Josd1
Genomic Map