siRNA Details
| Gene Symbol: Nfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290448.1 | NP_001277377.1 | 12 | 2178 | ||
| NM_001290449.1 | NP_001277378.1 | 12 | 2178 | |||
| NM_023739.3 | NP_076228.2 | 12 | 2141 | |||
| GenBank mRNA | AF223576.1 | - | 12 | 2105 | ||
| AK005038.2 | - | 12 | 2113 | |||
| AK033850.1 | - | 12 | 2072 | |||
| AK050749.1 | - | 12 | 2115 | |||
| AK144647.1 | - | 12 | 2127 | |||
| AK146108.1 | - | 12 | 2095 | |||
| AK155387.1 | - | 12 | 2093 | |||
| AK160013.1 | - | 12 | 2111 | |||
| BC053709.1 | - | 12 | 2076 | 30052746 | ||

Nfx1
Genomic Map