siRNA Details
| Gene Symbol: Fbxl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175206.4 | NP_780415.1 | 2 | 1497 | ||
| GenBank mRNA | AK035656.1 | - | 1 | 157 | ||
| AK052175.1 | - | 2 | 1484 | |||
| AK084845.1 | - | 2 | 1414 | |||
| AK085980.1 | - | 2 | 1441 | |||

Fbxl22
Genomic Map