siRNA Details
| Gene Symbol: Nudt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004444.1 | - | 2 | 198 | ||
| AK076500.1 | - | 2 | 185 | |||
| AK158058.1 | - | 2 | 260 | |||
| BC033921.1 | - | 2 | 170 | 5068809 | ||
| BY237462.1 | - | 2 | 309 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudt9| Gene Symbol: Nudt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004444.1 | - | 2 | 198 | ||
| AK076500.1 | - | 2 | 185 | |||
| AK158058.1 | - | 2 | 260 | |||
| BC033921.1 | - | 2 | 170 | 5068809 | ||
| BY237462.1 | - | 2 | 309 | |||