siRNA Details
| Gene Symbol: Nudt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004444.1 | - | 3 | 464 | ||
| AK076500.1 | - | 3 | 451 | |||
| AK151245.1 | - | 2 | 271 | |||
| AK158058.1 | - | 3 | 526 | |||
| AK206364.1 | - | 2 | 226 | |||
| BC033921.1 | - | 3 | 436 | 5068809 | ||

Nudt9
Genomic Map