siRNA Details
| Gene Symbol: Zdhhc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023740.2 | NP_076229.2 | 9 | 1198 | ||
| GenBank mRNA | AF176814.1 | - | 9 | 1171 | ||
| AK005255.1 | - | 8 | 930 | |||
| AK151505.1 | - | 9 | 1180 | |||
| AK168042.1 | - | 9 | 1173 | |||
| AK181099.1 | - | 2 | 162 | |||
| AK193583.1 | - | 2 | 162 | |||
| AK219891.1 | - | 2 | 162 | |||
| BC010835.1 | - | 9 | 1162 | 4239913 | ||

Zdhhc16
Genomic Map