siRNA Details
| Gene Symbol: Zdhhc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023740.2 | NP_076229.2 | 9, 10 | 1246 | ||
| GenBank mRNA | AF176814.1 | - | 9, 10 | 1219 | ||
| AK005255.1 | - | 8, 9 | 978 | |||
| AK151505.1 | - | 9, 10 | 1228 | |||
| AK168042.1 | - | 9, 10 | 1221 | |||
| AK206796.1 | - | 1, 2 | 22 | |||
| AK215392.1 | - | 1, 2 | 22 | |||
| BC010835.1 | - | 9, 10 | 1210 | 4239913 | ||

Zdhhc16
Genomic Map