siRNA Details
| Gene Symbol: Zdhhc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023740.2 | NP_076229.2 | 3 | 497 | ||
| GenBank mRNA | AF176814.1 | - | 3 | 470 | ||
| AK005255.1 | - | 1 | 93 | |||
| AK151505.1 | - | 3 | 479 | |||
| AK168042.1 | - | 3 | 472 | |||
| BC010835.1 | - | 3 | 461 | 4239913 | ||
| CK619221.1 | - | 3 | 484 | |||

Zdhhc16
Genomic Map