siRNA Details
| Gene Symbol: Stk40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145827.1 | NP_001139299.1 | 7 | 967 | ||
| NM_028800.3 | NP_083076.3 | 5 | 613 | |||
| GenBank mRNA | AB102946.1 | - | 5 | 444 | ||
| AK154780.1 | - | 7 | 958 | |||
| AK192521.1 | - | 1 | 57 | |||
| AY336057.1 | - | 5 | 598 | |||
| BC046981.1 | - | 5 | 572 | 6478265 | ||

Stk40
Genomic Map