siRNA Details
| Gene Symbol: Ctbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293672.1 | NP_001280601.1 | 2, 3 | 297 | ||
| NM_028836.4 | NP_083112.1 | 2, 3 | 297 | |||
| GenBank mRNA | AK008791.2 | - | 2, 3 | 272 | ||
| BC022594.1 | - | 1, 2 | 58 | |||
| DQ100384.1 | - | 2, 3 | 271 | |||

Ctbs
Genomic Map