siRNA Details
| Gene Symbol: Klrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033171.2 | NP_001028343.2 | 4, 5 | 1220 | ||
| GenBank mRNA | AK009417.1 | - | 4, 5 | 1221 | ||
| AK162933.1 | - | 5, 6 | 808 | |||
| AK169862.1 | - | 5, 6 | 764 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klrg2| Gene Symbol: Klrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033171.2 | NP_001028343.2 | 4, 5 | 1220 | ||
| GenBank mRNA | AK009417.1 | - | 4, 5 | 1221 | ||
| AK162933.1 | - | 5, 6 | 808 | |||
| AK169862.1 | - | 5, 6 | 764 | |||