siRNA Details
| Gene Symbol: Aven | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165935.1 | NP_001159407.1 | 2 | 121 | ||
| NM_028844.3 | NP_083120.2 | 3 | 451 | |||
| GenBank mRNA | AK005934.1 | - | 2 | 177 | ||
| AK006808.1 | - | 3 | 452 | |||
| BC100725.1 | - | 4 | 314 | 40045263 | ||
| BC100735.1 | - | 4 | 328 | |||
| DV646415.1 | - | 2 | 123 | |||

Aven
Genomic Map