siRNA Details
| Gene Symbol: Aven | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165935.1 | NP_001159407.1 | 2 | 162 | ||
| NM_028844.3 | NP_083120.2 | 3 | 492 | |||
| GenBank mRNA | AK006808.1 | - | 3 | 493 | ||
| AK046479.1 | - | 418 | ||||
| BC100725.1 | - | 4 | 355 | 40045263 | ||
| BC100735.1 | - | 4 | 369 | |||
| BX528932.1 | - | 1 | 22 | |||
| DV646415.1 | - | 2 | 164 | |||

Aven
Genomic Map