siRNA Details
| Gene Symbol: Usp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028846.5 | NP_083122.1 | 7 | 596 | ||
| GenBank mRNA | AK029890.1 | - | 493 | |||
| AK042612.1 | - | 7 | 513 | |||
| AK047242.1 | - | 7 | 515 | |||
| AK054279.1 | - | 7 | 539 | |||
| AK087412.1 | - | 7 | 507 | |||
| AK163663.1 | - | 7 | 506 | |||
| AK173083.1 | - | 7 | 498 | |||
| BC079674.1 | - | 485 | 30531994 | |||

Usp20
Genomic Map