siRNA Details
| Gene Symbol: Mageb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028847.1 | NP_083123.1 | 2 | 646 | ||
| GenBank mRNA | AK006807.1 | - | 2 | 689 | ||
| BC116773.1 | - | 2 | 646 | 40126092 | ||
| BC116775.1 | - | 2 | 646 | 40126094 | ||
| Gene Symbol: Gm14781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205268.1 | NP_001192197.1 | 2 | 691 | ||

Mageb5
Genomic Map